missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrrolidine 98.0+%, TCI America™
$50.05 - $229.10
Chemical Identifiers
| CAS | 120-94-5 |
|---|---|
| Molecular Formula | C5H11N |
| Molecular Weight (g/mol) | 85.15 |
| MDL Number | MFCD00003173 |
| InChI Key | AVFZOVWCLRSYKC-UHFFFAOYSA-N |
| Synonym | n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine |
| PubChem CID | 8454 |
| IUPAC Name | 1-methylpyrrolidine |
| SMILES | CN1CCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M041525ML
|
TCI America
M041525ML |
25 mL |
Each for $50.05
|
|
|||||
|
M0415100ML
|
TCI America
M0415100ML |
100 mL |
Each for $78.93
|
|
|||||
|
M0415500ML
|
TCI America
M0415500ML |
500 mL |
Each for $229.10
|
|
|||||
Chemical Identifiers
| 120-94-5 | |
| 85.15 | |
| AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
| 8454 | |
| CN1CCCC1 |
| C5H11N | |
| MFCD00003173 | |
| n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
| 1-methylpyrrolidine |
Specifications
| 120-94-5 | |
| 76°C | |
| MFCD00003173 | |
| 2924 | |
| AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
| 1-methylpyrrolidine | |
| 8454 | |
| ≥98.0% (GC,T) | |
| 1-Methylpyrrolidine |
| Yellow | |
| C5H11N | |
| 25 mL | |
| n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
| CN1CCCC1 | |
| 85.15 | |
| 85.15 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-6168
RTECSNumber : UY1420500
TSCA : Yes