missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrrolidine 98.0+%, TCI America™
Supplier: TCI America M0415100ML
Specifications
| 1-Methylpyrrolidine | |
| Yellow | |
| C5H11N | |
| 100 mL | |
| n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
| CN1CCCC1 | |
| 85.15 | |
| 85.15 | |
| Liquid |
| 120-94-5 | |
| 76°C | |
| MFCD00003173 | |
| 2924 | |
| AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
| 1-methylpyrrolidine | |
| 8454 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 120-94-5 | |
| 85.15 | |
| AVFZOVWCLRSYKC-UHFFFAOYSA-N | |
| 8454 | |
| CN1CCCC1 |
| C5H11N | |
| MFCD00003173 | |
| n-methylpyrrolidine, methylpyrrolidine, n-methyltetrahydropyrrole, pyrrolidine, 1-methyl, n-methyl pyrrolidine, 1-methyl-pyrrolidine, pyrrolidine, n-methyl, unii-06509tzu6c, methyl pyrrolidine | |
| 1-methylpyrrolidine |
Safety and Handling
EINECSNumber : (5)-6168
RTECSNumber : UY1420500
TSCA : Yes