missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole-2-carboxylic acid, 98%

Catalog No. AAL0590606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0590606 5 g
AAL0590603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0590606 Supplier Thermo Scientific Chemicals Supplier No. L0590606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6973-60-0 | C6H7NO2 | 125.127 g/mol

1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpyrrole-2-carboxylic acid is used for thermochemical and computational studies of 1-Methyl-2-pyrrolecarboxylic acid.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 6973-60-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00003088
InChI Key ILAOVOOZLVGAJF-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carboxylic acid, 1-methyl-2-pyrrolecarboxylic acid, n-methylpyrrole-2-carboxylic acid, 1h-pyrrole-2-carboxylic acid, 1-methyl, 1h-pyrrolecarboxylic acid, methyl, 1-methyl-1h-pyrrole-2-carboxylicacid, pubchem9181, acmc-209oar, ksc353i0t, 2-carboxy-1-methyl-1h-pyrrole
PubChem CID 81453
IUPAC Name 1-methylpyrrole-2-carboxylic acid
SMILES CN1C=CC=C1C(=O)O

Specifications

Melting Point 135°C to 138°C
Quantity 5 g
Beilstein 115703
Formula Weight 125.13
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-217-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only