missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrrole-2-carboxaldehyde, 98%

Catalog No. p-7081265
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1116309 10 g
AAA1116318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1116309 Supplier Thermo Scientific Chemicals Supplier No. A1116309
Only null left

CAS: 1192-58-1 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1192-58-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00003087
InChI Key OUKQTRFCDKSEPL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrrole-2-carbaldehyde, 1-methylpyrrole-2-carboxaldehyde, n-methylpyrrole-2-carboxaldehyde, n-methyl-2-pyrrolecarboxaldehyde, 1-methyl-1h-pyrrole-2-carboxaldehyde, 2-formyl-1-methylpyrrole, 1-methyl-2-pyrrolecarboxaldehyde, 1h-pyrrole-2-carboxaldehyde, 1-methyl, unii-m0hyh3d7sx, 1-methyl-2-formylpyrrole
PubChem CID 14504
IUPAC Name 1-methylpyrrole-2-carbaldehyde
SMILES CN1C=CC=C1C=O

Specifications

Density 1.074
Boiling Point 88°C to 90°C (22 mmHg)
Flash Point 72°C (161°F)
Refractive Index 1.561
Quantity 10 g
Beilstein 107811
Sensitivity Air and light sensitive
Formula Weight 109.13
Percent Purity 98%
Chemical Name or Material 1-Methylpyrrole-2-carboxaldehyde
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-755-0
RTECSNumber UX9350000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only