missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrazole 98.0+%, TCI America™
Supplier: TCI America M096925G
Specifications
| 1-Methylpyrazole | |
| Yellow | |
| C4H6N2 | |
| 25 g | |
| 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255 | |
| CN1C=CC=N1 | |
| 82.106 | |
| CHEBI:59025 | |
| ≥98.0% (GC) |
| 930-36-9 | |
| 127°C | |
| MFCD00144943 | |
| 1993 | |
| UQFQONCQIQEYPJ-UHFFFAOYSA-N | |
| 1-methylpyrazole | |
| 70255 | |
| 82.11 | |
| Liquid |
Chemical Identifiers
| 930-36-9 | |
| 82.106 | |
| UQFQONCQIQEYPJ-UHFFFAOYSA-N | |
| 70255 | |
| 1-methylpyrazole |
| C4H6N2 | |
| MFCD00144943 | |
| 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255 | |
| CHEBI:59025 | |
| CN1C=CC=N1 |
Safety and Handling
TSCA : No