missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrazole 98.0+%, TCI America™

Catalog No. p-7132469
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09695G 5 g
M096925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09695G Supplier TCI America Supplier No. M09695G
Only null left

Chemical Identifiers

CAS 930-36-9
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.106
MDL Number MFCD00144943
InChI Key UQFQONCQIQEYPJ-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole, n-methylpyrazole, 1h-pyrazole, 1-methyl, pyrazole, 1-methyl, methylpyrazole, 1-methyl pyrazole, 1-methypyrazole, n-methyl pyrazole, 1-methyl-pyrazole, zlchem 255
PubChem CID 70255
ChEBI CHEBI:59025
IUPAC Name 1-methylpyrazole
SMILES CN1C=CC=N1

Specifications

Color Yellow
Boiling Point 127°C
Quantity 5 g
UN Number 1993
Formula Weight 82.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrazole