missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpyrazole-4-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7133188
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23721G 1 g
M23725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23721G Supplier TCI America Supplier No. M23721G
Only null left

Chemical Identifiers

CAS 25016-11-9
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00460465
InChI Key MYFZXSOYJVWTBL-UHFFFAOYSA-N
Synonym 1-methyl-1h-pyrazole-4-carbaldehyde, 1-methyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde, 1-methyl, 4-formyl-1-methylpyrazole, 4-formyl-1-methyl-1h-pyrazole, 1-methylpyrazole-4-carboxaldehyde, 1-methylpyrazole-4-carboxyaldehyde, 1-methyl-1h-pyrazol-4-carbaldehyd, 1-methylpyrazole-4-aldehyde, acmc-1cs06
PubChem CID 573117
IUPAC Name 1-methylpyrazole-4-carbaldehyde
SMILES CN1C=C(C=N1)C=O

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 81°C
Quantity 1 g
Formula Weight 110.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylpyrazole-4-carboxaldehyde