accessibility menu, dialog, popup

UserName

CAS RN 25016-11-9

IUPAC Name: 1-methyl-1H-pyrazole-4-carbaldehyde
Synonyms: 1-methylpyrazole-4-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: MYFZXSOYJVWTBL-UHFFFAOYSA-N
SMILES: CN1C=C(C=O)C=N1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)