missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrazole-3-carboxylic Acid 97.0+%, TCI America™
$44.73 - $141.67
Chemical Identifiers
| CAS | 25016-20-0 |
|---|---|
| Molecular Formula | C5H6N2O2 |
| Molecular Weight (g/mol) | 126.12 |
| MDL Number | MFCD00464254 |
| InChI Key | YBFIKNNFQIBIQZ-UHFFFAOYSA-N |
| Synonym | 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid |
| PubChem CID | 573176 |
| ChEBI | CHEBI:74738 |
| IUPAC Name | 1-methyl-1H-pyrazole-3-carboxylic acid |
| SMILES | CN1C=CC(=N1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2642200MG
|
TCI America
M2642200MG |
200 mg |
Each for $44.73
|
|
|||||
|
M26421G
|
TCI America
M26421G |
1 g |
Each for $141.67
|
|
|||||
Chemical Identifiers
| 25016-20-0 | |
| 126.12 | |
| YBFIKNNFQIBIQZ-UHFFFAOYSA-N | |
| 573176 | |
| 1-methyl-1H-pyrazole-3-carboxylic acid |
| C5H6N2O2 | |
| MFCD00464254 | |
| 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid | |
| CHEBI:74738 | |
| CN1C=CC(=N1)C(O)=O |
Specifications
| 25016-20-0 | |
| White-Yellow | |
| MFCD00464254 | |
| 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid | |
| CN1C=CC(=N1)C(O)=O | |
| 126.12 | |
| CHEBI:74738 | |
| ≥97.0% (GC,T) | |
| 1-Methylpyrazole-3-carboxylic Acid |
| 154°C | |
| C5H6N2O2 | |
| 200 mg | |
| YBFIKNNFQIBIQZ-UHFFFAOYSA-N | |
| 1-methyl-1H-pyrazole-3-carboxylic acid | |
| 573176 | |
| 126.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No