missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpyrazole-3-carboxylic Acid 97.0+%, TCI America™
Supplier: TCI America M26421G
Specifications
| 1-Methylpyrazole-3-carboxylic Acid | |
| 154°C | |
| C5H6N2O2 | |
| 1 g | |
| YBFIKNNFQIBIQZ-UHFFFAOYSA-N | |
| 1-methyl-1H-pyrazole-3-carboxylic acid | |
| 573176 | |
| 126.12 | |
| Crystalline Powder |
| 25016-20-0 | |
| White-Yellow | |
| MFCD00464254 | |
| 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid | |
| CN1C=CC(=N1)C(O)=O | |
| 126.12 | |
| CHEBI:74738 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 25016-20-0 | |
| 126.12 | |
| YBFIKNNFQIBIQZ-UHFFFAOYSA-N | |
| 573176 | |
| 1-methyl-1H-pyrazole-3-carboxylic acid |
| C5H6N2O2 | |
| MFCD00464254 | |
| 1-methyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylic acid, 1-methyl, 3-carboxy-1-methyl-1h-pyrazole, zlchem 1152, acmc-209ggi, 1-methylpyrazolecarboxylic acid, 1-methyl-pyrazole-3-carboxylic acid, pyrazole-3-carboxylic acid, 1-methyl, 1h-pyrazole-3-carboxylicacid, 1-methyl, 1-methyl-1h-4-pyrazole-3-carboxylic acid | |
| CHEBI:74738 | |
| CN1C=CC(=N1)C(O)=O |
Safety and Handling
TSCA : No