missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine-2-methanol, 97%

CAS: 20845-34-5 | C7H15NO | 129.203 g/mol

Supplier:  Thermo Scientific Chemicals B2218714

 View more versions of this product

Catalog No. AAB2218714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Methylpiperidine-2-methanol
0.984
81°C (177°F)
1.483
25 g
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
CN1CCCCC1CO
129.203
129.2
20845-34-5
79°C to 80°C (7 mmHg)
C7H15NO
MFCD00006494
103612
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
(1-methylpiperidin-2-yl)methanol
89394
98%

Chemical Identifiers

20845-34-5
129.203
HXXJMMLIEYAFOZ-UHFFFAOYSA-N
89394
CN1CCCCC1CO
C7H15NO
MFCD00006494
1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
(1-methylpiperidin-2-yl)methanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 244-078-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only