missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine-2-methanol, 97%

Catalog No. p-7026271
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2218706 5 g
AAB2218714 25 g
AAB2218722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2218706 Supplier Thermo Scientific Chemicals Supplier No. B2218706

CAS: 20845-34-5 | C7H15NO | 129.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20845-34-5
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.203
MDL Number MFCD00006494
InChI Key HXXJMMLIEYAFOZ-UHFFFAOYSA-N
Synonym 1-methyl-2-piperidinemethanol, 1-methylpiperidin-2-yl methanol, 1-methylpiperidine-2-methanol, 2-piperidinemethanol, 1-methyl, 1-methyl-2-piperidyl methanol, 1-methyl-2-hydroxymethylpiperidine, 2-hydroxymethyl-1-methylpiperidine, 1-methyl-2-piperidyl methan-1-ol, pubchem8873, 1-methyl-2piperidinemethanol
PubChem CID 89394
IUPAC Name (1-methylpiperidin-2-yl)methanol
SMILES CN1CCCCC1CO

Specifications

Density 0.984
Boiling Point 79°C to 80°C (7 mmHg)
Flash Point 81°C (177°F)
Refractive Index 1.483
Quantity 5 g
Beilstein 103612
Formula Weight 129.2
Percent Purity 98%
Chemical Name or Material 1-Methylpiperidine-2-methanol
Hazard Category H227-H302-H314
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 244-078-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only