missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperazine, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019157
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019157 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019157 Supplier Merck Emd Millipore Supplier No. 145495MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

CAS 109-01-3
Vapor Density 3.5 (vs. air)
Density 0.903 g/mL (at 25°C)
Flash Point 39°C
Physical Form Neat
Linear Formula C5H12N2
MDL Number MFCD00005966
Boiling Point 138°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 100.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less