missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnicotinamide chloride, ≥97.5%, MilliporeSigma™ Supelco™

Catalog No. 111012274
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012274 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012274 Supplier Merck Emd Millipore Supplier No. M46271G
Only null left

1-Methylnicotinamide (MNA) is a primary metabolite of nicotinamide produced mainly by nicotinamide N-methyltransferase (NNMT) and exhibits antithrombotic and anti-inflammatory effects.

Featured Industries: Vitamins, Nutraceuticals and Natural Products

Specifications

Packaging Poly Bottle
CAS 1005-24-9
Physical Form Powder
Linear Formula C7H9N2 OCl
MDL Number MFCD00060042
Synonym N-Methylnicotinic acid amide chloride
Type Analytical Standard
Quantity 1 g
Formula Weight 172.61
Percent Purity ≥97.5%
Grade Analytical Standard
Show More Show Less