missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene, 96%

Catalog No. p-164864
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC127161000 100 g Glass bottle
AC127165000 500 g Glass bottle
AC127160025 2.5 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC127161000 Supplier Thermo Scientific Chemicals Supplier No. 127161000
Only null left

CAS: 90-12-0 | C11H10 | 142.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-12-0
Molecular Formula C11H10
Molecular Weight (g/mol) 142.2
MDL Number MFCD00004034
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
Synonym alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Specifications

CAS Min % >97
Melting Point -22.0°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 240.0°C to 243.0°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.6130 to 1.6160
Linear Formula C10H7CH3
Quantity 100 g
Beilstein 05, 566
Specific Gravity 1.02
Solubility Information Solubility in water: 26mg/l (25°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 142.2
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-Methylnaphthalene, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approve
EINECSNumber 201-966-8
RTECSNumber QJ9630000
TSCA TSCA

RUO – Research Use Only