missing translation for 'onlineSavingsMsg'
Learn More

1-Methylnaphthalene, 96%

Catalog No. AAA1532722
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1532722 100 g
AAA1532736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1532722 Supplier Thermo Scientific Chemicals Supplier No. A1532722

CAS: 90-12-0 | C11H10 | 142.201 g/mol

1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylnaphthalene is used as an important intermediate in organic synthesis. It is used as a starting material of surfactant, water reducer, dispersant and drugs. Further, it is employed in the preparation of 1-methylnaphthalene-d10 using deuterium oxide, sodium deuteroxide

Solubility
Miscible with water, alcohol, ethanol, ether and benzene.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, peroxides and oxygen.

Chemical Identifiers

CAS 90-12-0
Molecular Formula C11H10
Molecular Weight (g/mol) 142.201
MDL Number MFCD00004034
InChI Key QPUYECUOLPXSFR-UHFFFAOYSA-N
Synonym alpha-methylnaphthalene, naphthalene, 1-methyl, methylnaphthalene, naphthalene, methyl, polymethylnaphthalene, 1-methyl-naphthalene, alpha-methyl naphthalenes, methyl naphthalene, 1-methyl naphthalene, fema number 3193
PubChem CID 7002
ChEBI CHEBI:50717
IUPAC Name 1-methylnaphthalene
SMILES CC1=CC=CC2=CC=CC=C12

Specifications

Melting Point -22°C
Density 1.024
Boiling Point 240°C to 243°C
Flash Point 82°C (179°F)
Refractive Index 1.615
Quantity 100 g
UN Number UN3082
Beilstein 506793
Solubility Information Miscible with water,alcohol,ethanol,ether and benzene.
Formula Weight 142.2
Percent Purity 96%
Chemical Name or Material 1-Methylnaphthalene
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227-H315-H319
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 201-966-8
RTECSNumber QJ9630000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only