missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisoquinoline 98.0+%, TCI America™

Catalog No. p-7132092
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0298100MG 100 mg
M02981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0298100MG Supplier TCI America Supplier No. M0298100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1721-93-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006902
InChI Key PBYMYAJONQZORL-UHFFFAOYSA-N
Synonym isoquinaldine, isoquinoline, 1-methyl, 1-methyl-isoquinoline, unii-z96hox9rt9, isoquinoline, methyl, z96hox9rt9, methylisoquinoline, isoquinoline-3-carboxylicacid, 1-methyl isoquinoline, pubchem17034
PubChem CID 15592
IUPAC Name 1-methylisoquinoline
SMILES CC1=NC=CC2=CC=CC=C12

Specifications

Boiling Point 255°C
Quantity 100 mg
Formula Weight 143.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Methylisoquinoline