accessibility menu, dialog, popup

UserName

CAS RN 1721-93-3

IUPAC Name: 1-methylisoquinoline
Synonyms: isoquinoline, 1-methyl-
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: PBYMYAJONQZORL-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=CC=N1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)