missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisoquinoline, 97%

Catalog No. AAH2702903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2702903 1 g
AAH2702906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2702903 Supplier Thermo Scientific Chemicals Supplier No. H2702903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1721-93-3 | C10H9N | 143.189 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1721-93-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006902
InChI Key PBYMYAJONQZORL-UHFFFAOYSA-N
Synonym isoquinaldine, isoquinoline, 1-methyl, 1-methyl-isoquinoline, unii-z96hox9rt9, isoquinoline, methyl, z96hox9rt9, methylisoquinoline, isoquinoline-3-carboxylicacid, 1-methyl isoquinoline, pubchem17034
PubChem CID 15592
IUPAC Name 1-methylisoquinoline
SMILES CC1=NC=CC2=CC=CC=C12

Specifications

Melting Point 10°C to 12°C
Density 1.078
Boiling Point 126°C to 128°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.614
Quantity 1 g
Formula Weight 143.19
Percent Purity 97%
Chemical Name or Material 1-Methylisoquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-017-6
TSCA No

RUO – Research Use Only