missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisatin 98.0+%, TCI America™

Catalog No. M27065G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27065G 5 g
M27061G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27065G Supplier TCI America Supplier No. M27065G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2058-74-4
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005812
InChI Key VCYBVWFTGAZHGH-UHFFFAOYSA-N
Synonym n-methylisatin, 1-methylisatin, 1-methyl-1h-indole-2,3-dione, 1-methylindoline-2,3-dione, 1h-indole-2,3-dione, 1-methyl, 1-methyl-indole-2,3-dione, 1 methylisatine, n-methylindol-2,3-dione, 1-methyl-2,3-dihydro-1h-indole-2,3-dione, 1-methyl-2,3-indolinedione
PubChem CID 16358
IUPAC Name 1-methyl-2,3-dihydro-1H-indole-2,3-dione
SMILES CN1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 130°C
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methylisatin
RTECSNumber NL7939000
TSCA No