accessibility menu, dialog, popup

UserName

CAS RN 2058-74-4

IUPAC Name: 1-methyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: N-methylindol-2,3-dione
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: VCYBVWFTGAZHGH-UHFFFAOYSA-N
SMILES: CN1C(=O)C(=O)C2=CC=CC=C12
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (4)