missing translation for 'onlineSavingsMsg'
Learn More

1-Methylisatin, 97%

Catalog No. AAA1294203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1294203 1 g
AAA1294206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1294203 Supplier Thermo Scientific Chemicals Supplier No. A1294203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2058-74-4 | C9H7NO2 | 161.16 g/mol

1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylisatin is used as a reactant for stereoselective preparation of spirobicyclic and bis-spirotricyclic pyrazolidinones, for regioselective preparation of spirocyclic oxindole-butenolides, for synthesis of spiro-oxindoles, for preparation of unsymmetrical oxindoles, for stereoselective preparation of hydroxyloxindoles via Morita-Baylis-Hillman reaction, for preparation of pyridinecarboxylic acid [(oxo)indolylidene)hydrazide derivatives (Schiff base hydrazides) as antibacterial agents.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2058-74-4
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005812
InChI Key VCYBVWFTGAZHGH-UHFFFAOYSA-N
Synonym n-methylisatin, 1-methylisatin, 1-methyl-1h-indole-2,3-dione, 1-methylindoline-2,3-dione, 1h-indole-2,3-dione, 1-methyl, 1-methyl-indole-2,3-dione, 1 methylisatine, n-methylindol-2,3-dione, 1-methyl-2,3-dihydro-1h-indole-2,3-dione, 1-methyl-2,3-indolinedione
PubChem CID 16358
SMILES CN1C(=O)C(=O)C2=CC=CC=C12

Specifications

Melting Point 129°C to 133°C
Quantity 1 g
Beilstein 128280
Solubility Information Insoluble in water.
IUPAC Name 1-methylindole-2,3-dione
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 1-Methylisatin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-164-9
RTECSNumber NL7939000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only