missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylguanidine Hydrochloride 98.0+%, TCI America™
$86.64 - $221.39
Chemical Identifiers
| CAS | 21770-81-0 |
|---|---|
| Molecular Formula | C2H8ClN3 |
| Molecular Weight (g/mol) | 109.557 |
| MDL Number | MFCD00012576 |
| InChI Key | VJQCNCOGZPSOQZ-UHFFFAOYSA-N |
| PubChem CID | 146724 |
| IUPAC Name | 2-methylguanidine;hydrochloride |
| SMILES | CN=C(N)N.Cl |
Chemical Identifiers
| 21770-81-0 | |
| 109.557 | |
| VJQCNCOGZPSOQZ-UHFFFAOYSA-N | |
| 2-methylguanidine;hydrochloride |
| C2H8ClN3 | |
| MFCD00012576 | |
| 146724 | |
| CN=C(N)N.Cl |
Specifications
| 21770-81-0 | |
| C2H8ClN3 | |
| 5 g | |
| CN=C(N)N.Cl | |
| 109.557 | |
| 109.56 | |
| Crystalline Powder |
| White | |
| MFCD00012576 | |
| VJQCNCOGZPSOQZ-UHFFFAOYSA-N | |
| 2-methylguanidine;hydrochloride | |
| 146724 | |
| ≥98.0% (N,T) | |
| 1-Methylguanidine Hydrochloride |
Safety and Handling
RTECSNumber : MF3990000
TSCA : No