accessibility menu, dialog, popup

UserName

CAS RN 21770-81-0

ClH3CNH2NH2HN

CAS RN 21770-81-0

IUPAC Name: carbamimidoyl(methyl)azanium chloride
Synonyms: carbamimidoyl(methyl)azanium chloride
Molecular Weight (g/mol): 109.56
Molecular Formula: C2H8ClN3
InChi Key: VJQCNCOGZPSOQZ-UHFFFAOYSA-N
SMILES: [Cl-].C[NH2+]C(N)=N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results