missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylcyclopropane-1-carboxylic Acid 98.0+%, TCI America™
$275.87 - $937.62
Chemical Identifiers
| CAS | 6914-76-7 |
|---|---|
| Molecular Formula | C5H8O2 |
| Molecular Weight (g/mol) | 100.117 |
| MDL Number | MFCD00001290 |
| InChI Key | DIZKLZKLNKQFGB-UHFFFAOYSA-N |
| Synonym | 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa |
| PubChem CID | 81326 |
| IUPAC Name | 1-methylcyclopropane-1-carboxylic acid |
| SMILES | CC1(CC1)C(=O)O |
Chemical Identifiers
| 6914-76-7 | |
| 100.117 | |
| DIZKLZKLNKQFGB-UHFFFAOYSA-N | |
| 81326 | |
| CC1(CC1)C(=O)O |
| C5H8O2 | |
| MFCD00001290 | |
| 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa | |
| 1-methylcyclopropane-1-carboxylic acid |
Specifications
| 6914-76-7 | |
| 185°C | |
| MFCD00001290 | |
| 3261 | |
| DIZKLZKLNKQFGB-UHFFFAOYSA-N | |
| 1-methylcyclopropane-1-carboxylic acid | |
| 81326 | |
| ≥98.0% (GC,T) | |
| 1-Methylcyclopropane-1-carboxylic Acid |
| White-Yellow | |
| C5H8O2 | |
| 5 g | |
| 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa | |
| CC1(CC1)C(=O)O | |
| 100.117 | |
| 100.12 | |
| Crystalline Lumps |
Safety and Handling
TSCA : No