missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylcyclopropane-1-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M088525G
Specifications
| 1-Methylcyclopropane-1-carboxylic Acid | |
| White-Yellow | |
| C5H8O2 | |
| 25 g | |
| 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa | |
| CC1(CC1)C(=O)O | |
| 100.117 | |
| 100.12 | |
| Crystalline Lumps |
| 6914-76-7 | |
| 185°C | |
| MFCD00001290 | |
| 3261 | |
| DIZKLZKLNKQFGB-UHFFFAOYSA-N | |
| 1-methylcyclopropane-1-carboxylic acid | |
| 81326 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 6914-76-7 | |
| 100.117 | |
| DIZKLZKLNKQFGB-UHFFFAOYSA-N | |
| 81326 | |
| CC1(CC1)C(=O)O |
| C5H8O2 | |
| MFCD00001290 | |
| 1-methylcyclopropanecarboxylic acid, 1-methyl-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-methyl, cyclopropanecarboxylicacid, 1-methyl, pubchem16037, 1-methylcyclopropanoic acid, acmc-209o6g, d07fpq, 1-carboxy-1-methylcyclopropane, dizklzklnkqfgb-uhfffaoysa | |
| 1-methylcyclopropane-1-carboxylic acid |
Safety and Handling
TSCA : No