missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentene, 98%

Catalog No. p-7024343
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1881606 5 g
AAA1881614 25 g
AAA1881622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1881606 Supplier Thermo Scientific Chemicals Supplier No. A1881606
Only null left

CAS: 693-89-0 | C6H10 | 82.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 693-89-0
Molecular Formula C6H10
Molecular Weight (g/mol) 82.15
MDL Number MFCD00001397
InChI Key ATQUFXWBVZUTKO-UHFFFAOYSA-N
Synonym 1-methyl-1-cyclopentene, cyclopentene, 1-methyl, methylcyclopentene, cyclopentene, methyl, unii-v03m1dzc9i, 1-methylcyclopent-1-ene, v03m1dzc9i, methyl-cyclopentene, 1-methylcyclopentene-1, cyc1opentene,l-methyl
PubChem CID 12746
IUPAC Name 1-methylcyclopentene
SMILES CC1=CCCC1

Specifications

Melting Point -142°C
Density 0.779
Boiling Point 73°C to 75°C
Flash Point −17°C (1°F)
Refractive Index 1.431
Quantity 5 g
UN Number UN3295
Beilstein 1900640
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 1-Methylcyclopentene
Show More Show Less
Hazard Category H225-H304
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 211-762-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only