accessibility menu, dialog, popup

UserName

CH3

CAS RN 693-89-0

IUPAC Name: 1-methylcyclopent-1-ene
Synonyms: cyclopentene, 1-methyl-
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: ATQUFXWBVZUTKO-UHFFFAOYSA-N
SMILES: CC1=CCCC1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)