missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclohexanecarboxylic acid, 99%

Catalog No. AAB2171318
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2171318 50 g
AAB2171306 5 g
AAB2171314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2171318 Supplier Thermo Scientific Chemicals Supplier No. B2171318
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1123-25-7 | C8H14O2 | 142.198 g/mol

1-Methyl-1-cyclohexanecarboxylic acid was used as internal standard during the determination of valproic acid metabolites. Also been used as an anticonvulsant drug and causes maturation of murine neuroblastoma cells in vitro. Used in the synthesis of p38 map kinase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 1123-25-7
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00001463
InChI Key REHQLKUNRPCYEW-UHFFFAOYSA-N
Synonym 1-methylcyclohexanecarboxylic acid, 1-methyl-1-cyclohexanecarboxylic acid, 1-methylcyclohexylcarboxylic acid, 1-methylcyclohexanoic acid, cyclohexanecarboxylic acid, 1-methyl, 1-methyl-1-cyclohexanoic acid, 1-methyl-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, methyl, acmc-2099ec, 4-09-00-00041 beilstein handbook reference
PubChem CID 70744
IUPAC Name 1-methylcyclohexane-1-carboxylic acid
SMILES CC1(CCCCC1)C(=O)O

Specifications

Melting Point 36°C to 39°C
Boiling Point 236°C
Flash Point 100°C (212°F)
Quantity 50 g
Beilstein 1859220
Solubility Information Soluble in chloroform and methanol.
Formula Weight 142.2
Percent Purity 99%
Chemical Name or Material 1-Methylcyclohexanecarboxylic acid
EINECSNumber 214-371-3
RTECSNumber GU8585500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only