missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl L-Aspartate 98.0+%, TCI America™

Catalog No. p-7132882
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
M18591G 1 g
M18595G 5 g
2 options

Catalog No. M18591G

Supplier: TCI America M18591G

Only null left

Chemical Identifiers

CAS 17812-32-7
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00080289
InChI Key SWWBMHIMADRNIK-UHFFFAOYNA-N
Synonym h-asp-ome, 1-methyl l-aspartate, l-aspartic acid 1-methyl ester, s-3-amino-4-methoxy-4-oxobutanoic acid, 3s-3-amino-4-methoxy-4-oxobutanoic acid, l-aspartic acid methyl ester, pubchem13176, l-aspartic acid 1-methyl ester; 3-amino-4-methoxy-4-oxobutanoic acid; h-asp-ome
PubChem CID 6994682
IUPAC Name 3-amino-4-methoxy-4-oxobutanoic acid
SMILES COC(=O)C(N)CC(O)=O

Specifications

Melting Point 186°C
Color White
Quantity 1 g
Formula Weight 147.13
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl L-Aspartate