accessibility menu, dialog, popup

UserName

H3COOOOHNH2

CAS RN 17812-32-7

IUPAC Name: 3-amino-4-methoxy-4-oxobutanoic acid
Synonyms: 3-amino-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol): 147.13
Molecular Formula: C5H9NO4
InChi Key: SWWBMHIMADRNIK-UHFFFAOYNA-N
SMILES: COC(=O)C(N)CC(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)