missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic Acid 98.0+%, TCI America™
Supplier: TCI America M27531G
Specifications
| 1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic Acid | |
| 123°C | |
| C6H5F3N2O2 | |
| 1 g | |
| CN1C(=C(C=N1)C(=O)O)C(F)(F)F | |
| 194.113 | |
| 194.11 | |
| Crystalline Powder |
| 119083-00-0 | |
| White-Yellow | |
| MFCD04110182 | |
| VAKOSNKAXYJZRG-UHFFFAOYSA-N | |
| 1-methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid | |
| 2807771 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 119083-00-0 | |
| 194.113 | |
| VAKOSNKAXYJZRG-UHFFFAOYSA-N | |
| 1-methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid |
| C6H5F3N2O2 | |
| MFCD04110182 | |
| 2807771 | |
| CN1C(=C(C=N1)C(=O)O)C(F)(F)F |
Safety and Handling
TSCA : No