accessibility menu, dialog, popup

UserName

CAS RN 119083-00-0

CH3NNOHOFFF

CAS RN 119083-00-0

IUPAC Name: 1-methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid
Synonyms: 1-methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Molecular Weight (g/mol): 194.11
Molecular Formula: C6H5F3N2O2
InChi Key: VAKOSNKAXYJZRG-UHFFFAOYSA-N
SMILES: CN1N=CC(C(O)=O)=C1C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results