missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine 98.0+%, TCI America™

Catalog No. p-7133085
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21975G 5 g
M219725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21975G Supplier TCI America Supplier No. M21975G
Only null left

Chemical Identifiers

CAS 1629218-99-0
Molecular Formula C15H30N4
Molecular Weight (g/mol) 266.43
MDL Number MFCD20257913
InChI Key VVNRVKMOTTTYKS-UHFFFAOYSA-N
PubChem CID 53384439
IUPAC Name 4-(4-methylpiperazin-1-yl)-1,4'-bipiperidine
SMILES CN1CCN(CC1)C1CCN(CC1)C1CCNCC1

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 5 g
Synonym 1-Methyl-4-(1,4′C-bipiperidin-4-yl)piperazine
Formula Weight 266.43
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-4-[1-(4-piperidyl)-4-piperidyl]piperazine