accessibility menu, dialog, popup

UserName

CH3NNNNH

CAS RN 1629218-99-0

IUPAC Name: 4-(4-methylpiperazin-1-yl)-1,4'-bipiperidine
Synonyms: 4-(4-methylpiperazin-1-yl)-1,4'-bipiperidine
Molecular Weight (g/mol): 266.43
Molecular Formula: C15H30N4
InChi Key: VVNRVKMOTTTYKS-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1CCN(CC1)C1CCNCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results