missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-phenylthiourea 97.0+%, TCI America™

Catalog No. M27861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27861G 1 g
M27865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27861G Supplier TCI America Supplier No. M27861G

Chemical Identifiers

CAS 2724-69-8
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.242
MDL Number MFCD00022130
InChI Key IGEQFPWPMCIYDF-UHFFFAOYSA-N
PubChem CID 698294
IUPAC Name 1-methyl-3-phenylthiourea
SMILES CNC(=S)NC1=CC=CC=C1

Specifications

Melting Point 112°C
Color White-Yellow
Quantity 1 g
UN Number 2811
Formula Weight 166.24
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3-phenylthiourea
RTECSNumber YT8750000
TSCA Yes