missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-p-tolyltriazene 98.0+%, TCI America™

Catalog No. p-7132278
Change view
Click to view available options
Quantity:
1 g
25 g
Catalog No. Quantity
M06411G 1 g
M064125G 25 g
2 options

Catalog No. M06411G

Supplier: TCI America M06411G

Only null left

Chemical Identifiers

CAS 21124-13-0
Molecular Formula C8H11N3
Molecular Weight (g/mol) 149.197
MDL Number MFCD00008282
InChI Key DNGJVDGPCGXBFF-UHFFFAOYSA-N
PubChem CID 30492
IUPAC Name 4-methyl-N-(methyldiazenyl)aniline
SMILES CC1=CC=C(C=C1)NN=NC

Specifications

Melting Point 81°C
Color Yellow
Quantity 1 g
UN Number 1325
Formula Weight 149.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3-p-tolyltriazene [for Esterification]
RTECSNumber XY2785500
TSCA Yes