missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™

$234.87 - $861.19

Chemical Identifiers

CAS 403842-84-2
Molecular Formula C13H23F3N2O3S
Molecular Weight (g/mol) 344.39
MDL Number MFCD06411717
InChI Key BCVMAYRUJBXMAP-UHFFFAOYSA-M
Synonym 1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet
PubChem CID 16212746
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24405G
View Documents
TCI America
M24405G
5 g
Each for $234.87
Only null left
 
M244025G
View Documents
TCI America
M244025G
25 g
Each for $861.19
Only null left
 
Description

Description

Specifications

Chemical Identifiers

403842-84-2
344.39
BCVMAYRUJBXMAP-UHFFFAOYSA-M
16212746
[O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
C13H23F3N2O3S
MFCD06411717
1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet
3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate

Specifications

403842-84-2
C13H23F3N2O3S
5 g
BCVMAYRUJBXMAP-UHFFFAOYSA-M
3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate
16212746
≥98.0% (HPLC,N)
1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate
Yellow
MFCD06411717
1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet
[O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1
344.39
344.39
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'