missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate 98.0+%, TCI America™
Supplier: TCI America M244025G
Specifications
| 1-Methyl-3-n-octylimidazolium Trifluoromethanesulfonate | |
| Yellow | |
| MFCD06411717 | |
| 1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet | |
| [O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1 | |
| 344.39 | |
| 344.39 | |
| Liquid |
| 403842-84-2 | |
| C13H23F3N2O3S | |
| 25 g | |
| BCVMAYRUJBXMAP-UHFFFAOYSA-M | |
| 3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate | |
| 16212746 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 403842-84-2 | |
| 344.39 | |
| BCVMAYRUJBXMAP-UHFFFAOYSA-M | |
| 16212746 | |
| [O-]S(=O)(=O)C(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1 |
| C13H23F3N2O3S | |
| MFCD06411717 | |
| 1-methyl-3-octylimidazolium trifluoromethanesulfonate, 1-methyl-3-n-octylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium trifluoromethanesulfonate, 1-octyl-3-methylimidazolium triflate, dsstox_cid_27922, dsstox_rid_82675, dsstox_gsid_47946, 1-methyl-3-octylimidazolium triflate, 1-methyl-3-n-octylimidazolium triflate, 1-methyl-3-octylimidazolium trifluoromet | |
| 3-methyl-1-octyl-1H-imidazol-3-ium trifluoromethanesulfonate |
Safety and Handling
TSCA : No