missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Hexafluorophosphate 98.0+%, TCI America™

Catalog No. p-7132998
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M20635G 5 g
M206325G 25 g
2 options

Catalog No. M20635G

Supplier: TCI America M20635G

Only null left

Chemical Identifiers

CAS 304680-36-2
Molecular Formula C12H23F6N2P
Molecular Weight (g/mol) 340.29
MDL Number MFCD03427617
InChI Key GRCIJNHHTXBJAK-UHFFFAOYSA-N
Synonym 1-methyl-3-n-octylimidazolium hexafluorophosphate, 3-methyl-1-octylimidazolium hexafluorophosphate, 1-methyl-3-octylimidazolium hexafluorophosphate, 1-octyl-3-methylimidazolium hexafluorophosphate, 1-methyl-3-n-octylimidazoliumhexafluorophosphate, acmc-20aj2z, dsstox_cid_27928, dsstox_rid_82681, dsstox_gsid_47952, ksc222c5l
PubChem CID 2734243
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 340.29
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1-Methyl-3-n-octylimidazolium Hexafluorophosphate