missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-n-octylimidazolium Chloride 98.0+%, TCI America™
$98.18 - $292.62
Chemical Identifiers
| CAS | 64697-40-1 |
|---|---|
| Molecular Formula | C12H23ClN2 |
| Molecular Weight (g/mol) | 230.78 |
| MDL Number | MFCD03095432 |
| InChI Key | OXFBEEDAZHXDHB-UHFFFAOYSA-M |
| Synonym | 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j |
| PubChem CID | 2734223 |
| ChEBI | CHEBI:61342 |
| IUPAC Name | 3-methyl-1-octyl-1H-imidazol-3-ium chloride |
| SMILES | [Cl-].CCCCCCCCN1C=C[N+](C)=C1 |
Chemical Identifiers
| 64697-40-1 | |
| 230.78 | |
| OXFBEEDAZHXDHB-UHFFFAOYSA-M | |
| 2734223 | |
| 3-methyl-1-octyl-1H-imidazol-3-ium chloride |
| C12H23ClN2 | |
| MFCD03095432 | |
| 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j | |
| CHEBI:61342 | |
| [Cl-].CCCCCCCCN1C=C[N+](C)=C1 |
Specifications
| 64697-40-1 | |
| C12H23ClN2 | |
| 5 g | |
| OXFBEEDAZHXDHB-UHFFFAOYSA-M | |
| 3-methyl-1-octyl-1H-imidazol-3-ium chloride | |
| 2734223 | |
| 230.78 | |
| Liquid |
| 12°C | |
| MFCD03095432 | |
| 1-methyl-3-n-octylimidazolium chloride, 1-methyl-3-octylimidazolium chloride, 1-octyl-3-methylimidazolium chloride, 3-methyl-1-octylimidazolium chloride, omimcl, acmc-20aj1j, dsstox_cid_27849, dsstox_rid_82602, dsstox_gsid_47873, ksc491c7j | |
| [Cl-].CCCCCCCCN1C=C[N+](C)=C1 | |
| 230.78 | |
| CHEBI:61342 | |
| ≥98.0% (HPLC,T) | |
| 1-Methyl-3-n-octylimidazolium Chloride |
Safety and Handling
TSCA : No