missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%
CAS: 178631-04-4 | C14H23F6N3O4S2 | 475.465 g/mol
$147.77 - $1309.86
Chemical Identifiers
| CAS | 178631-04-4 |
|---|---|
| Molecular Formula | C14H23F6N3O4S2 |
| Molecular Weight (g/mol) | 475.465 |
| MDL Number | MFCD08458946 |
| InChI Key | LECQXINNQGHJBM-UHFFFAOYSA-N |
| Synonym | 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide |
| PubChem CID | 12184591 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium |
| SMILES | CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH3535206
|
Thermo Scientific Chemicals
H3535206 |
5 g |
Each for $147.77
|
|
|||||
|
AAH3535214
|
Thermo Scientific Chemicals
H3535214 |
25 g |
Each for $481.93
|
|
|||||
|
AAH3535222
|
Thermo Scientific Chemicals
H3535222 |
100 g |
Each for $1,309.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 178631-04-4 | |
| 475.465 | |
| LECQXINNQGHJBM-UHFFFAOYSA-N | |
| 12184591 | |
| CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
| C14H23F6N3O4S2 | |
| MFCD08458946 | |
| 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium |
Specifications
| 178631-04-4 | |
| MFCD08458946 | |
| 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide | |
| CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 475.465 | |
| 475.46 | |
| 1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide |
| C14H23F6N3O4S2 | |
| 5 g | |
| LECQXINNQGHJBM-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium | |
| 12184591 | |
| 99% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only