accessibility menu, dialog, popup

UserName

H3CNNCH3FFFSOONSOOFFF

CAS RN 178631-04-4

IUPAC Name: 3-methyl-1-octyl-1H-imidazol-3-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: 1-octyl-3-methylimidazolium; bistriflylimide anion
Molecular Weight (g/mol): 475.47
Molecular Formula: C14H23F6N3O4S2
InChi Key: LECQXINNQGHJBM-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C=C[N+](C)=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)