missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide, 99%

Catalog No. AAH3535206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3535206 5 g
AAH3535214 25 g
AAH3535222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3535206 Supplier Thermo Scientific Chemicals Supplier No. H3535206

CAS: 178631-04-4 | C14H23F6N3O4S2 | 475.465 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 178631-04-4
Molecular Formula C14H23F6N3O4S2
Molecular Weight (g/mol) 475.465
MDL Number MFCD08458946
InChI Key LECQXINNQGHJBM-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide
PubChem CID 12184591
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium
SMILES CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Formula Weight 475.46
Percent Purity 99%
Chemical Name or Material 1-Methyl-3-n-octylimidazolium bis(trifluoromethylsulfonyl)imide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only