missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulfonyl)imide 97.0+%, TCI America™

$167.76 - $559.22

Chemical Identifiers

CAS 178631-04-4
Molecular Formula C14H23F6N3O4S2
Molecular Weight (g/mol) 475.465
MDL Number MFCD08458946
InChI Key LECQXINNQGHJBM-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide
PubChem CID 12184591
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium
SMILES CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M30395G
View Documents
TCI America
M30395G
5 g
Each for $167.76
Only null left
 
M303925G
View Documents
TCI America
M303925G
25 g
Each for $559.22
Only null left
 
Description

Description

Specifications

Chemical Identifiers

178631-04-4
475.465
LECQXINNQGHJBM-UHFFFAOYSA-N
12184591
CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
C14H23F6N3O4S2
MFCD08458946
1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide
bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium

Specifications

178631-04-4
MFCD08458946
1-methyl-3-octylimidazolium bis trifluoromethylsulfonyl imide, dsstox_cid_27931, dsstox_rid_82684, dsstox_gsid_47955, 1-octyl-3-methylimidazolium; bistriflylimide anion, 1-n-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-octyl-3-methylimidazolium bis trifluoromethylsulfonyl imide, 1-methyl-3-n-octylimidazolium bis trifluoromethanesulfonyl imide
CCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
475.465
475.47
Liquid
C14H23F6N3O4S2
5 g
LECQXINNQGHJBM-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;1-methyl-3-octylimidazol-1-ium
12184591
≥97.0% (HPLC)
1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulfonyl)imide
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'