missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-Pyrrolidone, MilliporeSigma™
Supplier: Technidata MX13904
Specifications
| 1-Methyl-2-Pyrrolidone | |
| 872-50-4 | |
| Glass Bottle | |
| C5H9NO | |
| 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
| CN1CCCC1=O | |
| 99.133 | |
| CHEBI:7307 |
| OmniPure™ | |
| 500 mL | |
| Liquid | |
| (CH3)-NC(:O)CH2CH2CH2- | |
| SECXISVLQFMRJM-UHFFFAOYSA-N | |
| 1-methylpyrrolidin-2-one | |
| 13387 | |
| 99.13 |
Chemical Identifiers
| 872-50-4 | |
| 99.133 | |
| 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl | |
| CHEBI:7307 | |
| CN1CCCC1=O |
| C5H9NO | |
| SECXISVLQFMRJM-UHFFFAOYSA-N | |
| 13387 | |
| 1-methylpyrrolidin-2-one |
Safety and Handling
Recommended Storage : Room Temperature