missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidone, ACS, 99%, Spectrum™ Chemical

Catalog No. p-7165875
Change view
Click to view available options
Quantity:
20 L
200 L
Packaging:
Drum
Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608120 20 L Steel Can
18608121 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608120 Supplier Spectrum Chemical Mfg Cor Supplier No. M128020LT
Only null left

Chemical Identifiers

CAS 872-50-4
Molecular Weight (g/mol) 99.14

Specifications

CAS Max % 1
Boiling Point 202°C
Quantity 20 L
Packaging Steel Can
Water 0.0005
Percent Purity 0.99
Grade Reagent
Chemical Name or Material 1-Methyl-2-pyrrolidone

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.