missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidone, 99.0% min., ACS, MilliporeSigma™

Catalog No. MMX13951
Change view
Click to view available options
Quantity:
3 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX13951 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMX13951 Supplier Technidata Supplier No. MX13951

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Assay Percent Range 99.0% min.
Linear Formula (CH3)-NC(:O)CH2CH2CH2-
Quantity 3 kg
Formula Weight 99.13
Grade ACS
Physical Form Liquid
Chemical Name or Material 1-Methyl-2-pyrrolidone, For ACS analysis
Recommended Storage Room Temperature