missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, Spectrophotometric Grade, 99.5+%

Catalog No. AA39176K2
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39176K2 1 L
AA39176M1 4 x 1 L
AA39176K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39176K2 Supplier Thermo Scientific Chemicals Supplier No. 039176K2
Get Up to $1,800 in Chemicals with Purchase
View all promotions

For Spectrophotometry/ Chromatography | CAS: 872-50-4 | C5H9NO | 99.13 g/mol

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection. It is useful for the chemical functionalization of graphene sheets by solvothermal reduction of graphene oxide. It is also useful for pharmaceutical and different industrial applications due to its broad solvency and low volatility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Evaporation residue 6ppm, H2O 0.05%

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Melting Point °C to 24°C
Color Colorless
Appearance Colorless liquid
Density 1.033
Quantity 1 L
Flash Point 86°C (187°F)
Ignition Point 270°C (518°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.4705
Vapor Pressure 0.29 hPa
Beilstein 106420
Boiling Point 204°C
Solubility Information Miscible with water,lower alcohols,ketones,ethyl acetate,castor oil,chloroform and benzene. Moderately miscible with aliphatic hydrocarbons.
Formula Weight 99.13
Grade Spectrophotometry
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
Hazard Category H227-H315-H319-H335-H360D-H373
Precautionary Statement P201-P202-P210-P235-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
EINECSNumber 212-828-1
RTECSNumber UY5790000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only