missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, ACS Reagent, ≥99.0%, Solstice

Catalog No. 60046887
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046887 1 L Glass Bottle
60046888 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046887 Supplier Solstice Supplier No. 4437781L

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Density 1.028g/mL at 25°C
Quantity 1 L
Explosion Limit 9.5%
Ignition Point 518°C
Packaging Glass Bottle
Vapor Pressure 0.29mmHg (20°C), 0.99mmHg (40°C)
Beilstein 106420
Formula Weight 99.13g/mol
Percent Purity ≥99.0%
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Hazard Statement P201-P280-P305 + P351 + P338-P308 + P313
RTECSNumber 212-828-1